Novus Biologicals
Manufacturer Code:NBP276371
Catalog # NBP276371
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence between amino acids 623 and 673 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Polyclonal Antibody
Protein Aliases: Protein slit-like 2; Protein slit-like 2 SLITL2 slit-like 2 slit-like 2 (Drosophila) vasorin; slit-like 2; Vasorin
Gene Aliases: SLITL2; UNQ314/PRO357/PRO1282; VASN
UniProt ID: (Human) Q6EMK4
Entrez Gene ID: (Human) 114990
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.