Novus Biologicals
Manufacturer Code:NB10098886
Catalog # NB10098886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide as a part of the 1t cytoplasmic loop of human TRPV1 conjugated to an immunogenic carrier protein was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Capsaicin receptor; Capsaicin receptor Osm-9-like TRP channel 1 OTRPC1 transient receptor potential cation channel subfamily V member 1 transient receptor potential cation channel subfamily V member 1 transient receptor potential vanilloid 1a transient receptor potential vanilloid 1b TrpV1 Vanilloid receptor 1 vanilloid receptor subtype 1 VR1 VR1DKFZp434K0220; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TrpV1; Vanilloid receptor 1; vanilloid receptor subtype 1
Gene Aliases: TRPV1; VR1
UniProt ID: (Human) Q8NER1
Entrez Gene ID: (Human) 7442
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.