R&D Systems
Manufacturer Code:AF4815SP
Catalog # AF4815SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Vang-like Protein 2/VANGL2 Gln241-Val521 Accession # Q9ULK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1215Loop-tail protein 1 homolog loop-tail-associated protein LPP1 LTAP LTAPVan Gogh-like protein 2 MGC119403 MGC119404 STB1 STB1Strabismus 1 STBM STBM1 Strabismus vang (van gogh Drosophila)-like 2 vang van gogh-like 2 (Drosophila) Vang like Protein 2 VANGL2 vang-like 2 (van gogh Drosophila) vang-like protein 2; Loop-tail protein 1 homolog; loop-tail-associated protein; Strabismus 1; Van Gogh-like protein 2; vang-like 2 (van gogh, Drosophila); Vang-like protein 2
Gene Aliases: KIAA1215; LPP1; LTAP; STB1; STBM; STBM1; VANGL2
UniProt ID: (Human) Q9ULK5
Entrez Gene ID: (Human) 57216
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.