Abnova
Manufacturer Code:PAB23176
Catalog # 89-124-875
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human VWA2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant VWA2.
For Research Use Only
Protein Aliases: A domain-containing protein similar to matrilin and collagen; AMACO; CCSP-2; colon cancer diagnostic marker; Colon cancer secreted protein 2; MGC157755; von Willebrand factor A domain-containing protein 2
Gene Aliases: AMACO; CCSP-2; NET42; VWA2
UniProt ID: (Human) Q5GFL6
Entrez Gene ID: (Human) 340706
Molecular Function: extracellular matrix glycoprotein receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.