Proteintech
Manufacturer Code:187141AP150UL
Catalog # 50-173-2205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A domain-containing protein similar to matrilin and collagen; AMACO; AMACO CCSP 2 NET42 VWA2; CCSP-2; colon cancer diagnostic marker; Colon cancer secreted protein 2; von Willebrand factor A domain-containing protein 2
Gene Aliases: AMACO; CCSP-2; NET42; VWA2
UniProt ID: (Human) Q5GFL6
Entrez Gene ID: (Human) 340706
Molecular Function:
extracellular matrix glycoprotein
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.