Proteintech
Manufacturer Code:663981IG150UL
Catalog # 50-173-6826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2C8 |
Immunogen | VTN Fusion Protein Ag8443 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: complement S-protein; epibolin; Glycoprotein 66; S protein Serum spreading factor V75 vitronectin VN VNT VTN; S-protein; serum spreading factor; Serum-spreading factor; somatomedin B; Somatomedin-B; V75; Vitronectin; Vitronectin V10 subunit; Vitronectin V65 subunit; VN
Gene Aliases: V75; VN; VNT; VTN
UniProt ID: (Human) P04004, (Pig) P48819, (Rabbit) P22458
Entrez Gene ID: (Human) 7448, (Pig) 397192, (Rabbit) 100009128
Molecular Function:
cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.