Invitrogen
Manufacturer Code:PA572877
Catalog # PIPA572877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VTI1b antibody for a 17 amino acid peptide near the center of human VTI1b. The immunogen is within amino acids 150 - 200 of VTI1b. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A novel SNARE; Soluble N-ethylmaleimide-sensitive factor-attachment protein receptor; vesicle transport through interaction with t-SNAREs 1B; vesicle transport through interaction with t-SNAREs 1B homolog; Vesicle transport through interaction with t-SNAREs homolog 1B; Vesicle transport v-SNARE protein Vti1-like 1; vesicle-associated soluble NSF attachment protein receptor; Vti1-rp1; VTI1B vesicle transport through interaction with t-SNAREs homolog 1B (yeast) HGNC:17793 VTI1 VTI1L VTI2 v-SNARE vesicle transport through interaction with t-SNAREs 1B vesicle-associated soluble NSF attachment protein receptor
Gene Aliases: AU015348; GES30; MVti1b; SNARE; v-SNARE; VTI1; VTI1-LIKE; Vti1-rp1; VTI1B; VTI1L; VTI1L1; VTI2
UniProt ID: (Human) Q9UEU0
Entrez Gene ID: (Human) 10490, (Rat) 100359512, (Mouse) 53612
Molecular Function: SNARE protein membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.