Novus Biologicals
Manufacturer Code:NBP293359100UL
Catalog # NB028600
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human VTI1A (NP_660207.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MVti1 vesicle transport through interaction with t-SNAREs homolog 1A vesicle transport through interaction with t-SNAREs homolog 1A (yeast); SNARE Vti1a-beta protein; Vesicle transport through interaction with t-SNAREs homolog 1A; Vesicle transport v-SNARE protein Vti1-like 2; Vti1-rp2
Gene Aliases: MMDS3; MVti1; Vti1-rp2; VTI1A; VTI1RP2
UniProt ID: (Human) Q96AJ9
Entrez Gene ID: (Human) 143187
Molecular Function:
SNARE protein
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.