Abnova
Manufacturer Code:PAB9842
Catalog # 89-134-447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to amino acids 264-361 of human VSX2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Sheep polyclonal antibody raised against partial recombinant VSX2.
For Research Use Only
Protein Aliases: ceh-10 homeo domain containing homolog; Ceh-10 homeodomain-containing homolog; CHX10; Homeobox protein CHX10; HOX10; MCOP2; MCOPCB3; RET1; Visual system homeobox 2
Gene Aliases: CHX10; HOX10; MCOP2; MCOPCB3; RET1; VSX2
UniProt ID: (Human) P58304
Entrez Gene ID: (Human) 338917
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.