Proteintech
Manufacturer Code:258251AP150UL
Catalog # 50-173-5088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VSX2 Fusion Protein Ag23035 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C. elegans ceh-10 homeo domain containing homolog; ceh-10 homeo domain containing homolog; Ceh-10 homeodomain-containing homolog; CHX10 Homeobox protein CHX10 HOX10 MCOP2 MCOPCB3 RET1 visual system homeobox 2 VSX2; Homeobox protein CHX10; ocular retardation; Visual system homeobox 2
Gene Aliases: CHX10; Hox-10; HOX10; MCOP2; MCOPCB3; or; RET1; VSX2
UniProt ID: (Human) P58304, (Mouse) Q61412
Entrez Gene ID: (Human) 338917, (Mouse) 12677
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.