Invitrogen
Manufacturer Code:PA597243
Catalog # PIPA597243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-170 of human VSX1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeodomain protein RINX; Retinal inner nuclear layer homeobox protein; Transcription factor VSX1; Visual system homeobox 1
Gene Aliases: CAASDS; KTCN; KTCN1; PPCD; PPCD1; PPD; RINX; VSX1
UniProt ID: (Human) Q9NZR4
Entrez Gene ID: (Human) 30813
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.