Invitrogen
Manufacturer Code:PA5100803
Catalog # PIPA5100803
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human VSX1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CAASDS CHX10-like Homeodomain protein RINX KTCN KTCN1 PPCD PPCD1 PPD Retinal inner nuclear layer homeobox protein RINX Transcription factor VSX1 visual system homeobox 1 visual system homeobox 1 homolog (zebrafish) Vsx1; Homeodomain protein RINX; Retinal inner nuclear layer homeobox protein; Transcription factor VSX1; Visual system homeobox 1
Gene Aliases: CAASDS; CHX10-like; KTCN; KTCN1; PPCD; PPCD1; PPD; RINX; VSX1
UniProt ID: (Human) Q9NZR4, (Mouse) Q91V10
Entrez Gene ID: (Human) 30813, (Mouse) 114889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.