OriGene
Manufacturer Code:UM800035CF
Catalog # 50-168-5201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB116 |
Immunogen | Human recombit protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Hippocalcin-like protein 3; HLP3; HLP3 HPCAL3 HUVISL1 VILIP VILIP-1; VILIP; visinin-like 1; Visinin-like protein 1; VLP-1
Gene Aliases: HLP3; HPCAL3; HUVISL1; VILIP; VILIP-1; VISL1; VSNL1
UniProt ID: (Human) P62760
Entrez Gene ID: (Human) 7447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.