Invitrogen
Manufacturer Code:25575280
Catalog # 25-575-280
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | NLA14 |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: complement receptor of the immunoglobulin superfamily; CRIg Z39Ig complement receptor of the immunoglobulin superfamily Ig superfamily protein protein Z39Ig; V-set and immunoglobulin domain-containing protein 4
Gene Aliases: A530061A11; BC025105; CRIg; Z39IG
Entrez Gene ID: (Mouse) 278180
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.