Invitrogen
Manufacturer Code:17575741
Catalog # 17-575-741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | JAV4 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: complement receptor of the immunoglobulin superfamily; CRIg Z39Ig complement receptor of the immunoglobulin superfamily Ig superfamily protein protein Z39Ig; Ig superfamily protein; Protein Z39Ig; V-set and immunoglobulin domain-containing protein 4
Gene Aliases: CRIg; UNQ317/PRO362; VSIG4; Z39IG
UniProt ID: (Human) Q9Y279
Entrez Gene ID: (Human) 11326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.