Novus Biologicals
Manufacturer Code:NBP169631
Catalog # NBP169631
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to VSIG4(V-set and immunoglobulin domain containing 4) The peptide sequence was selected from the N terminal of VSIG4. Peptide sequence VPGDVSLQLSTLEMDDRSHYTCEVTWQTPDGNQVVRDKITELRVQKLSVS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complement receptor of the immunoglobulin superfamily; CRIg Ig superfamily protein Protein Z39Ig V-set and immunoglobulin domain containing 4 V-set and immunoglobulin domain-containing protein 4 Z39IGcomplement receptor of the immunoglobulin superfamily; Ig superfamily protein; Protein Z39Ig; V-set and immunoglobulin domain-containing protein 4
Gene Aliases: CRIg; UNQ317/PRO362; VSIG4; Z39IG
UniProt ID: (Human) Q9Y279
Entrez Gene ID: (Human) 11326
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.