Invitrogen
Manufacturer Code:PA570923
Catalog # PIPA570923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human VRTN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: vertebrae development homolog (pig); Vertnin
Gene Aliases: C14orf115; vertnin; VRTN
UniProt ID: (Pig) E1CHH8, (Human) Q9H8Y1
Entrez Gene ID: (Dog) 490782, (Pig) 100157734, (Human) 55237
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.