Invitrogen
Manufacturer Code:MA1828
Catalog # PIMA1828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F6 |
Immunogen | Recombit fusion protein GST-VRK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Serine/threonine-protein kinase 51PK; Serine/threonine-protein kinase VRK1; Vaccinia related kinase 1 MGC117401 MGC138280 Serine/threonine-protein kinase VRK1 Vaccinia-related kinase 1; vaccinia virus B1R-related kinase 1; Vaccinia-related kinase 1
Gene Aliases: 51PK; PCH1; PCH1A; VRK1
UniProt ID: (Human) Q99986, (Mouse) Q80X41
Entrez Gene ID: (Human) 7443, (Mouse) 22367
Molecular Function:
kinase
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.