Invitrogen
Manufacturer Code:MA524456
Catalog # PIMA524456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C8 |
Immunogen | KIAA0804 (NP&lowbar001009921 1329 a.a. &bsim 1428 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: vacuolar protein sorting 8 homolog; Vacuolar protein sorting-associated protein 8 homolog; VPS8 CORVET complex subunit; VPS8 KIAA0804
Gene Aliases: KIAA0804; VPS8
UniProt ID: (Human) Q8N3P4
Entrez Gene ID: (Human) 23355
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.