Proteintech
Manufacturer Code:151431AP150UL
Catalog # 50-173-0017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS72 Fusion Protein Ag7424 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CFL1 Protein YL 1 Swc2 TCFL1 Transcription factor like 1 VPS72 YL 1 YL1; Protein YL-1; Transcription factor-like 1; transformation suppressor gene YL-1; Vacuolar protein sorting-associated protein 72 homolog
Gene Aliases: CFL1; Swc2; TCFL1; VPS72; YL-1; YL1
UniProt ID: (Human) A6PW55, (Mouse) Q62481
Entrez Gene ID: (Human) 6944, (Rat) 310661, (Mouse) 21427
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.