Abnova
Manufacturer Code:H00006944K
Catalog # 89-209-765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human VPS72 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human VPS72 peptide using ARM Technology.
For Research Use Only
Protein Aliases: CFL1 Swc2 TCFL1 YL-1 YL1; Protein YL-1; Transcription factor-like 1; transformation suppressor gene YL-1; Vacuolar protein sorting-associated protein 72 homolog
Gene Aliases: CFL1; Swc2; TCFL1; VPS72; YL-1; YL1
UniProt ID: (Human) A6PW55
Entrez Gene ID: (Human) 6944
Molecular Function:
DNA-directed RNA polymerase
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.