Proteintech
Manufacturer Code:133271AP150UL
Catalog # 50-172-8654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS54 Fusion Protein Ag4123 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCC8 hVps54L SLP 8p Tumor antigen HOM HCC 8 Tumor antigen SLP 8p VPS54 VPS54L; Hepatocellular carcinoma protein 8; protein phosphatase 1, regulatory subunit 164; Tumor antigen HOM-HCC-8; Tumor antigen SLP-8p; Tumor antigen SLP-8p homolog; vacuolar protein sorting 54 homolog; Vacuolar protein sorting-associated protein 54; VPS54 GARP complex subunit; Vps54-like protein
Gene Aliases: HCC8; hVps54L; mSLP8; PPP1R164; SLP-8p; VPS54; VPS54L; Vsp54; WR
UniProt ID: (Human) Q9P1Q0, (Rat) Q9JMK8, (Mouse) Q5SPW0
Entrez Gene ID: (Human) 51542, (Rat) 286932, (Mouse) 245944
Molecular Function:
nucleic acid binding
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.