Invitrogen
Manufacturer Code:PA555079
Catalog # PIPA555079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human VPS53 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PP13624 HCCS1 hVps53L pp13624 vacuolar protein sorting-associated protein 53 homolog; SAC2 suppressor of actin mutations 2-like protein; Vacuolar protein sorting-associated protein 52 homolog; Vacuolar protein sorting-associated protein 53 homolog; VPS53 GARP complex subunit
Gene Aliases: 2010002A08Rik; 2310040I21Rik; 3100002B05Rik; Are1; HCCS1; hVps53L; PCH2E; PP13624; Sacm2l; Vps52; VPS53
UniProt ID: (Human) Q5VIR6, (Rat) O55166, (Mouse) Q8CCB4
Entrez Gene ID: (Human) 55275, (Rat) 25218, (Mouse) 68299
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.