Proteintech
Manufacturer Code:128241AP150UL
Catalog # 50-172-8277
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS53 Fusion Protein Ag3518 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HCCS1 hVps53L pp13624 VPS53; Vacuolar protein sorting-associated protein 53 homolog; VPS53 GARP complex subunit
Gene Aliases: 2010002A08Rik; 2310040I21Rik; 3100002B05Rik; HCCS1; hVps53L; PCH2E; PP13624; RGD1311391; VPS53
UniProt ID: (Human) Q5VIR6, (Mouse) Q8CCB4
Entrez Gene ID: (Human) 55275, (Mouse) 68299, (Rat) 287535
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.