Novus Biologicals
Manufacturer Code:NBP297979IR
Catalog # NB050370
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human VPS52 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARE1 dJ1033B10.5 DKFZp547I194 SAC2 SAC2 suppressor of actin mutations 2-like (yeast) SAC2 suppressor of actin mutations 2-like protein SACM2LRP5-1033B10 vacuolar protein sorting 52 (yeast) vacuolar protein sorting 52 homolog (S. cerevisiae) vacuolar protein sorting-associated protein 52 homolog; SAC2 suppressor of actin mutations 2-like protein; vacuolar protein sorting 52 homolog; Vacuolar protein sorting-associated protein 52 homolog; VPS52 GARP complex subunit
Gene Aliases: ARE1; dJ1033B10.5; SAC2; SACM2L; VPS52
UniProt ID: (Human) Q8N1B4
Entrez Gene ID: (Human) 6293
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.