Invitrogen
Manufacturer Code:PA575945
Catalog # PIPA575945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human VPS4B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell migration-inducing 1; Cell migration-inducing gene 1 protein; Protein SKD1; Suppressor of K(+) transport growth defect 1; suppressor of K+ transport defect 1; vacuolar protein sorting 4B; Vacuolar protein sorting-associated protein 4B; Vacuolar protein sorting-associated protein 4B Cell migration-inducing gene 1 protein Suppressor of K(+) transport growth defect 1 Protein SKD1 VPS4B SKD1 VPS42 MIG1
Gene Aliases: 8030489C12Rik; MIG1; SKD1; SKD1B; VPS4-2; VPS42; VPS4B
UniProt ID: (Human) O75351, (Mouse) P46467
Entrez Gene ID: (Human) 9525, (Mouse) 20479, (Rat) 360834
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.