Novus Biologicals
Manufacturer Code:NBP119118
Catalog # NBP119118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human vacuolar protein sorting 4 homolog B using the numbering given in entry NP_004860.2 (GeneID 9525). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cell migration-inducing 1; Cell migration-inducing gene 1 protein; Cell migration-inducing gene 1 protein Protein SKD1 SKD1B SKD1cell migration-inducing 1 Suppressor of K(+) transport growth defect 1 suppressor of K+ transport defect 1 vacuolar protein sorting 4 homolog B (S. cerevisiae) vacuolar protein sorting 4B vacuolar protein sorting 4B (yeast) vacuolar protein sorting-associated protein 4B VPS42 VPS4-2; Protein SKD1; Suppressor of K(+) transport growth defect 1; suppressor of K+ transport defect 1; vacuolar protein sorting 4B; Vacuolar protein sorting-associated protein 4B
Gene Aliases: MIG1; SKD1; SKD1B; VPS4-2; VPS42; VPS4B
UniProt ID: (Human) O75351
Entrez Gene ID: (Human) 9525
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.