Invitrogen
Manufacturer Code:PA577129
Catalog # PIPA577129
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human VPS4A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS4; Protein SKD2; SKD1-homolog; vacuolar protein sorting 4a; vacuolar protein sorting factor 4A; vacuolar protein sorting protein 4a; Vacuolar protein sorting-associated protein 4A; Vacuolar protein sorting-associated protein 4A Protein SKD2 VPS4-1 Hvps4 VPS4A VPS4; vacuolar sorting protein 4; VPS4-1; VPS4A
Gene Aliases: 4930589C15Rik; AI325971; AW553189; SKD1; SKD1A; SKD2; VPS4; VPS4-1; vps4-A; VPS4A
UniProt ID: (Human) Q9UN37, (Rat) Q793F9, (Mouse) Q8VEJ9
Entrez Gene ID: (Human) 27183, (Rat) 246772, (Mouse) 116733
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.