Invitrogen
Manufacturer Code:PA554176
Catalog # PIPA554176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human VPS41 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HVPS41 HVSP41 hVps41p S53 vacuolar assembly protein 41 vacuolar protein sorting-associated protein 41 homolog; S53; vacuolar assembly protein 41; vacuolar protein sorting 41 homolog; Vacuolar protein sorting-associated protein 41 homolog; VPS41 HOPS complex subunit
Gene Aliases: HVPS41; hVps41p; HVSP41; VPS41
UniProt ID: (Human) P49754
Entrez Gene ID: (Human) 27072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.