Invitrogen
Manufacturer Code:PA542303
Catalog # PIPA542303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human VPS41 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mVAM2; S53; vacuolar assembly protein 41; vacuolar protein sorting 41 homolog; vacuolar protein sorting 41 homolog (S. cerevisiae); Vacuolar protein sorting-associated protein 41 homolog; Vam2; VAM2 homolog; VPS41 HOPS complex subunit; VPS41, HOPS complex subunit
Gene Aliases: AI317346; fa01f11; HVPS41; hVps41p; HVSP41; mVam2; RGD1560511; si:ch211-146l10.2; Vam2; VPS41; wu:fa01f11
UniProt ID: (Human) P49754, (Mouse) Q5KU39
Entrez Gene ID: (Horse) 100062843, (Human) 27072, (Rabbit) 100343722, (Bovine) 528654, (Dog) 475876, (Mouse) 218035, (Rat) 306991, (Guinea pig) 100719226, (Zebrafish) 563215
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.