Novus Biologicals
Manufacturer Code:NB1002425
Catalog # NB1002425
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DHLKKDSQNKTLLK corresponding to internal region according to NP_055211.2 NP_542198.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: HVPS41 hVps41p HVSP41 S53 vacuolar assembly protein 41 vacuolar protein sorting 41 (yeast homolog) vacuolar protein sorting 41 (yeast) vacuolar protein sorting 41 homolog (S. cerevisiae) vacuolar protein sorting-associated protein 41 homolog; S53; vacuolar assembly protein 41; vacuolar protein sorting 41 homolog; Vacuolar protein sorting-associated protein 41 homolog; VPS41 HOPS complex subunit
Gene Aliases: HVPS41; hVps41p; HVSP41; VPS41
UniProt ID: (Human) P49754
Entrez Gene ID: (Human) 27072
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.