Novus Biologicals
Manufacturer Code:NBP176535
Catalog # NBP176535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human VPS39. The immunogen is located within amino acids 710 - 760 of VPS39. Amino Acid Squence: HKHYDRNKDGNKDVYLS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVam6p; hVam6p KIAA0770FLJ21681 TLP TRAP1-like protein vacuolar protein sorting 39 (yeast) vacuolar protein sorting 39 homolog (S. cerevisiae) vam6/Vps39-like protein VAM6FLJ46546; TRAP1-like protein; vacuolar protein sorting 39 homolog; Vam6/Vps39-like protein; VPS39 HOPS complex subunit
Gene Aliases: hVam6p; KIAA0770; TLP; VAM6; VPS39
UniProt ID: (Human) Q96JC1
Entrez Gene ID: (Human) 23339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.