Abnova
Manufacturer Code:PAB19373
Catalog # 89-120-907
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids near C-terminus of human VPS39. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of VPS39.
For Research Use Only
Protein Aliases: FLJ21681; FLJ46546; hVam6p; KIAA0770; TLP; TRAP1-like protein; vacuolar protein sorting 39 homolog; VAM6; Vam6/Vps39-like protein; VPS39 HOPS complex subunit
Gene Aliases: hVam6p; KIAA0770; TLP; VAM6; VPS39
UniProt ID: (Human) Q96JC1
Entrez Gene ID: (Human) 23339
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.