Proteintech
Manufacturer Code:162191AP150UL
Catalog # 50-173-0802
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS39 Fusion Protein Ag9267 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVam6p; hVam6p KIAA0770 TLP VAM6 Vam6/Vps39 like protein VPS39; TRAP1-like protein; vacuolar protein sorting 39 homolog; Vam6/Vps39-like protein; VPS39 HOPS complex subunit
Gene Aliases: A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; Pldn; TLP; VAM6; Vam6P; VPS39
UniProt ID: (Human) Q96JC1, (Mouse) Q8R5L3
Entrez Gene ID: (Human) 23339, (Mouse) 269338
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.