Proteintech
Manufacturer Code:663271IG150UL
Catalog # 50-173-6759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1F9A4 |
Immunogen | VPS37A Fusion Protein Ag19108 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ESCRT I complex subunit VPS37A HCRP1 hVps37A PQBP2 VPS37A; ESCRT-I complex subunit VPS37A; Hepatocellular carcinoma-related protein 1; hVps37A; polyglutamine binding protein 2; vacuolar protein sorting 37 homolog A; Vacuolar protein sorting-associated protein 37A
Gene Aliases: HCRP1; PQBP2; SPG53; VPS37A
UniProt ID: (Human) Q8NEZ2
Entrez Gene ID: (Human) 137492
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.