Invitrogen
Manufacturer Code:PA5116118
Catalog # PIPA5116118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human VPS36 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700010A24Rik 2210415M20Rik 2810408E15Rik C13orf9 CGI-145 Eap45 ELL-associated protein of 45 kDa ESCRT-II complex subunit VPS36 RGD1309754 vacuolar protein sorting 36 (yeast) vacuolar protein sorting 36 homolog vacuolar protein sorting 36 homolog (S. cerevisiae) vacuolar protein-sorting-associated protein 36 Vps36; ELL-associated protein of 45 kDa; ESCRT-II complex subunit VPS36; Vacuolar protein-sorting-associated protein 36
Gene Aliases: 1700010A24Rik; 2210415M20Rik; 2810408E15Rik; C13orf9; CGI-145; EAP45; RGD1309754; VPS36
UniProt ID: (Human) Q86VN1, (Mouse) Q9CVA3
Entrez Gene ID: (Human) 51028, (Rat) 290851, (Mouse) 70160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.