Novus Biologicals
Manufacturer Code:NBP1034300.025ML
Catalog # NB018264
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the internal part of mouse Vacuolar protein-sorting-associated protein 36 (VPS36) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C13orf9 CGI-145 DKFZp781E0871 Eap45 EAP45chromosome 13 open reading frame 9 ELL-associated protein of 45 kDa ELL-associated protein 45 kDa ESCRT-II complex subunit VPS36 vacuolar protein sorting 36 homolog (S. cerevisiae) vacuolar protein sorting 36 homolog (yeast) vacuolar protein-sorting-associated protein 36; ELL-associated protein of 45 kDa; ESCRT-II complex subunit VPS36; Vacuolar protein-sorting-associated protein 36
Gene Aliases: C13orf9; CGI-145; EAP45; VPS36
UniProt ID: (Human) Q86VN1
Entrez Gene ID: (Human) 51028
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.