Invitrogen
Manufacturer Code:MA534647
Catalog # PIMA534647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB33-82 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: AI647796 DKFZp434E1211 DKFZp434P1672 FLJ10752 FLJ13588 FLJ20388 hVPS35 Maternal-embryonic 3 Mem3 PARK17 TCCCTA00141 Vacuolar protein sorting-associated protein 35 Vesicle protein sorting 35 Vps35; hVPS35; Maternal-embryonic 3; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35
Gene Aliases: MEM3; PARK17; TCCCTA00141; VPS35
UniProt ID: (Human) Q96QK1
Entrez Gene ID: (Human) 55737
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.