Novus Biologicals
Manufacturer Code:NBP278827
Catalog # NB002994
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 11H10 |
Immunogen | Full length recombinant human VSP35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp434P1672 FLJ10752 FLJ13588 FLJ20388 hVPS35 Maternal-embryonic 3 MEM3DKFZp434E1211 vacuolar protein sorting 35 (yeast homolog) vacuolar protein sorting 35 (yeast) vacuolar protein sorting 35 homolog (S. cerevisiae) vacuolar protein sorting-associated protein 35 Vesicle protein sorting 35; hVPS35; Maternal-embryonic 3; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35
Gene Aliases: MEM3; PARK17; TCCCTA00141; VPS35
UniProt ID: (Human) Q96QK1
Entrez Gene ID: (Human) 55737
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.