Invitrogen
Manufacturer Code:PA530654
Catalog # PIPA530654
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 311 of Human VPS35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS35; Maternal embryonic message 3; Maternal-embryonic 3; MEM3 PARK17 hVPS35 maternal-embryonic 3 vacuolar protein sorting-associated protein 35; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35
Gene Aliases: AI647796; MEM3; PARK17; TCCCTA00141; VPS35
UniProt ID: (Human) Q96QK1, (Mouse) Q9EQH3
Entrez Gene ID: (Human) 55737, (Rat) 25479, (Mouse) 65114
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.