Invitrogen
Manufacturer Code:PA518248
Catalog # PIPA518248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SPESEGPIYEGLIL) corresponding to the C-terminus amino acids of VPS35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS35; Maternal embryonic message 3; Maternal-embryonic 3; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35; VPS35 MEM3 FLJ10752 FLJ13588 FLJ20388 DKFZp434E1211 DKFZp434P1672 vacuolar protein sorting 35 (yeast) maternal-embryonic 3 vacuolar protein sorting 35 homolog (S. cerevisiae) vacuolar protein sorting 35
Gene Aliases: AI647796; MEM3; PARK17; TCCCTA00141; VPS35
UniProt ID: (Human) Q96QK1, (Mouse) Q9EQH3
Entrez Gene ID: (Human) 55737, (Rat) 25479, (Mouse) 65114
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.