Novus Biologicals
Manufacturer Code:NBP220876
Catalog # NBP220876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human VPS35. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434P1672 FLJ10752 FLJ13588 FLJ20388 hVPS35 Maternal-embryonic 3 MEM3DKFZp434E1211 vacuolar protein sorting 35 (yeast homolog) vacuolar protein sorting 35 (yeast) vacuolar protein sorting 35 homolog (S. cerevisiae) vacuolar protein sorting-associated protein 35 Vesicle protein sorting 35; hVPS35; Maternal-embryonic 3; vacuolar protein sorting 35 homolog; Vacuolar protein sorting-associated protein 35; Vesicle protein sorting 35
Gene Aliases: MEM3; PARK17; TCCCTA00141; VPS35
UniProt ID: (Human) Q96QK1
Entrez Gene ID: (Human) 55737
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.