Proteintech
Manufacturer Code:124521AP150UL
Catalog # 50-172-7991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS34 (C terminal) Fusion Protein Ag3110 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: catalytic phosphatidylinositol 3-kinase 3; hVps34; hVps34 PI 3 Kinase Class 3 PI 3-kinase p100 PI3K p100 PI3K type 3 PIK3C3 PtdIns 3 kinase type 3 VPS34; Phosphatidylinositol 3-kinase catalytic subunit type 3; Phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI-3K Vps34p; PI3-kinase type 3; PI3K type 3; PtdIns-3-kinase type 3
Gene Aliases: 5330434F23Rik; hVps34; PIK3C3; VPS34
UniProt ID: (Human) Q8NEB9, (Rat) O88763, (Mouse) Q6PF93
Entrez Gene ID: (Human) 5289, (Rat) 65052, (Mouse) 225326
Molecular Function:
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.