Proteintech
Manufacturer Code:121951AP150UL
Catalog # 50-554-964
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS33B Fusion Protein Ag2833 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS33B; hVPS33B VPS33B; r-vps33b; vacuolar protein sorting 33 homolog B; vacuolar protein sorting 33-like protein B; vacuolar protein sorting 33B; vacuolar protein sorting homolog r-vps33b; Vacuolar protein sorting-associated protein 33B; vacuolar sorting protein 33b; vps33b
Gene Aliases: VPS33B; zgc:110803
UniProt ID: (Human) Q9H267, (Mouse) P59016, (Rat) Q63616, (Zebrafish) Q58EN8
Entrez Gene ID: (Human) 26276, (Mouse) 233405, (Rat) 64060, (Zebrafish) 541534
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.