Invitrogen
Manufacturer Code:PA545268
Catalog # PIPA545268
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human VPS33A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: buff; hVPS33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33 homolog A (S. cerevisiae); vacuolar protein sorting 33A; vacuolar protein sorting protein 33a; Vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A CORVET/HOPS core subunit; VPS33A, CORVET/HOPS core subunit
Gene Aliases: 3830421M04Rik; AI503300; AW048546; AW554476; Bf; Buff; im:7136893; r-Vps33a; VPS33A; wu:fc08g01; wu:fc26c10
UniProt ID: (Human) Q96AX1, (Mouse) Q9D2N9
Entrez Gene ID: (Rabbit) 100344276, (Horse) 100058750, (Human) 65082, (Bovine) 510197, (Dog) 477462, (Mouse) 77573, (Rat) 65081, (Guinea pig) 100724252, (Zebrafish) 448942
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.