Invitrogen
Manufacturer Code:PA531456
Catalog # PIPA531456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 365 and 596 of Human VPS33A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hVPS33A; hVPS33A vacuolar protein sorting 33A vacuolar protein sorting-associated protein 33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33A; Vacuolar protein sorting-associated protein 33A; VPS33A CORVET/HOPS core subunit
Gene Aliases: VPS33A
UniProt ID: (Human) Q96AX1
Entrez Gene ID: (Human) 65082
Molecular Function: membrane traffic protein membrane trafficking regulatory protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.