Novus Biologicals
Manufacturer Code:NBP220872
Catalog # NBP220872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human VPS33A. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ22395 FLJ23187 hVPS33A vacuolar protein sorting 33 homolog A (S. cerevisiae) vacuolar protein sorting 33A vacuolar protein sorting 33A (yeast) vacuolar protein sorting-associated protein 33A; hVPS33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33A; Vacuolar protein sorting-associated protein 33A; VPS33A CORVET/HOPS core subunit
Gene Aliases: VPS33A
UniProt ID: (Human) Q96AX1
Entrez Gene ID: (Human) 65082
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.