Proteintech
Manufacturer Code:168961AP150UL
Catalog # 50-173-1243
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | VPS33A Fusion Protein Ag10422 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: buff; hVPS33A; hVPS33A VPS33A; vacuolar protein sorting 33 homolog A; vacuolar protein sorting 33A; vacuolar protein sorting protein 33a; Vacuolar protein sorting-associated protein 33A; vacuolar sorting protein 33a; VPS33A CORVET/HOPS core subunit
Gene Aliases: 3830421M04Rik; AI503300; AW048546; AW554476; Bf; Buff; r-Vps33a; VPS33A
UniProt ID: (Human) Q96AX1, (Mouse) Q9D2N9
Entrez Gene ID: (Human) 65082, (Mouse) 77573, (Rat) 65081
Molecular Function:
membrane traffic protein
membrane trafficking regulatory protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.