OriGene
Manufacturer Code:CF505691
Catalog # 50-166-3838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1A8 |
Immunogen | Full length human recombit protein of human VPS28 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Caspase-activated DNase inhibitor that interacts with ASK1; CIIA; ESCRT-I complex subunit VPS28; H-Vps28; vacuolar protein sorting 28 homolog; vacuolar protein sorting 28-like protein; Vacuolar protein sorting-associated protein 28 homolog; VPS28 ESCRT-I subunit; VPS28, ESCRT-I subunit; yeast class E protein Vps28p homolog
Gene Aliases: 1110014J03Rik; CIIA; D730005C08Rik; VPS28
UniProt ID: (Human) Q9UK41, (Mouse) Q9D1C8
Entrez Gene ID: (Dog) 607677, (Human) 51160, (Mouse) 66914, (Rat) 300052
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.