Invitrogen
Manufacturer Code:PA530553
Catalog # PIPA530553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 4 and 221 of Human VPS28 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ESCRT-I complex subunit VPS28; ESCRT-I complex subunit VPS28 H-Vps28 vacuolar protein sorting-associated protein 28 homolog yeast class E protein Vps28p homolog; H-Vps28; vacuolar protein sorting 28 homolog; vacuolar protein sorting 28-like protein; Vacuolar protein sorting-associated protein 28 homolog; yeast class E protein Vps28p homolog
Gene Aliases: VPS28
UniProt ID: (Human) Q9UK41
Entrez Gene ID: (Human) 51160
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.